MilliporeSigma
  • Improved protein arrays for quantitative systems analysis of the dynamics of signaling pathway interactions.
Improved protein arrays for quantitative systems analysis of the dynamics of signaling pathway interactions.
Proteome science (2011-09-16)
Xiaoyu Wang, Ying Dong, Ameena J Jiwani, Yonglong Zou, Johanne Pastor, Makoto Kuro-O, Amyn A Habib, Minzi Ruan, David A Boothman, Chin-Rang Yang
ABSTRACT
An improved version of quantitative protein array platform utilizing linear Quantum dot signaling for systematically measuring protein levels and phosphorylation states is presented. The signals are amplified linearly by a confocal laser Quantum dot scanner resulting in ~1000-fold more sensitivity than traditional Western blots, but are not linear by the enzyme-based amplification. Software is developed to facilitate the quantitative readouts of signaling network activities. Kinetics of EGFRvIII mutant signaling was analyzed to quantify cross-talks between EGFR and other signaling pathways.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Millipore
ReBlot Plus Mild Antibody Stripping Solution, 10x