MilliporeSigma
  • Cleavage by MMP-13 renders VWF unable to bind to collagen but increases its platelet reactivity.
Cleavage by MMP-13 renders VWF unable to bind to collagen but increases its platelet reactivity.
Journal of thrombosis and haemostasis : JTH (2020-01-03)
Joanna-Marie Howes, Vera Knäuper, Jean-Daniel Malcor, Richard W Farndale
ABSTRACT
Atherosclerotic plaque rupture and subsequent thrombosis underpin thrombotic syndromes. Under inflammatory conditions in the unstable plaque, perturbed endothelial cells secrete von Willebrand Factor (VWF) which, via its interaction with GpIbα, enables platelet rolling across and adherence to the damaged endothelium. Following plaque rupture, VWF and platelets are exposed to subendothelial collagen, which supports stable platelet adhesion, activation, and aggregation. Plaque-derived matrix metalloproteinase (MMP)-13 is also released into the surrounding lumen where it may interact with VWF, collagen, and platelets. We sought to discover whether MMP-13 can cleave VWF and whether this might regulate its interaction with both collagen and platelets. We have used platelet adhesion assays and whole blood flow experiments to assess the effects of VWF cleavage by MMP-13 on platelet adhesion and thrombus formation. Unlike the shear-dependent cleavage of VWF by a disintegrin and metalloprotease with thrombospondin motif member 13 (ADAMTS13), MMP-13 is able to cleave VWF under static conditions. Following cleavage by MMP-13, immobilized VWF cannot bind to collagen but interacts more strongly with platelets, supporting slower platelet rolling in whole blood under shear. Compared with intact VWF, the interaction of cleaved VWF with platelets results in greater GpIbα upregulation and P-selectin expression, and the thrombi formed on cleaved VWF-collagen co-coatings are larger and more contractile than platelet aggregates on intact VWF-collagen co-coatings or on collagen alone. Our data suggest a VWF-mediated role for MMP-13 in the recruitment of platelets to the site of vascular injury and may provide new insights into the association of MMP-13 in atherothrombotic and stroke pathologies.
Sigma-Aldrich
Anti-Sheep IgG (whole molecule)−FITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution