MilliporeSigma
  • Inhibitors of histone acetyltransferases KAT6A/B induce senescence and arrest tumour growth.
Inhibitors of histone acetyltransferases KAT6A/B induce senescence and arrest tumour growth.
Nature (2018-08-03)
Jonathan B Baell, David J Leaver, Stefan J Hermans, Gemma L Kelly, Margs S Brennan, Natalie L Downer, Nghi Nguyen, Johannes Wichmann, Helen M McRae, Yuqing Yang, Ben Cleary, H Rachel Lagiakos, Stephen Mieruszynski, Guido Pacini, Hannah K Vanyai, Maria I Bergamasco, Rose E May, Bethany K Davey, Kimberly J Morgan, Andrew J Sealey, Beinan Wang, Natasha Zamudio, Stephen Wilcox, Alexandra L Garnham, Bilal N Sheikh, Brandon J Aubrey, Karen Doggett, Matthew C Chung, Melanie de Silva, John Bentley, Pat Pilling, Meghan Hattarki, Olan Dolezal, Matthew L Dennis, Hendrik Falk, Bin Ren, Susan A Charman, Karen L White, Jai Rautela, Andrea Newbold, Edwin D Hawkins, Ricky W Johnstone, Nicholas D Huntington, Thomas S Peat, Joan K Heath, Andreas Strasser, Michael W Parker, Gordon K Smyth, Ian P Street, Brendon J Monahan, Anne K Voss, Tim Thomas
ABSTRACT
Acetylation of histones by lysine acetyltransferases (KATs) is essential for chromatin organization and function1. Among the genes coding for the MYST family of KATs (KAT5-KAT8) are the oncogenes KAT6A (also known as MOZ) and KAT6B (also known as MORF and QKF)2,3. KAT6A has essential roles in normal haematopoietic stem cells4-6 and is the target of recurrent chromosomal translocations, causing acute myeloid leukaemia7,8. Similarly, chromosomal translocations in KAT6B have been identified in diverse cancers8. KAT6A suppresses cellular senescence through the regulation of suppressors of the CDKN2A locus9,10, a function that requires its KAT activity10. Loss of one allele of KAT6A extends the median survival of mice with MYC-induced lymphoma from 105 to 413 days11. These findings suggest that inhibition of KAT6A and KAT6B may provide a therapeutic benefit in cancer. Here we present highly potent, selective inhibitors of KAT6A and KAT6B, denoted WM-8014 and WM-1119. Biochemical and structural studies demonstrate that these compounds are reversible competitors of acetyl coenzyme A and inhibit MYST-catalysed histone acetylation. WM-8014 and WM-1119 induce cell cycle exit and cellular senescence without causing DNA damage. Senescence is INK4A/ARF-dependent and is accompanied by changes in gene expression that are typical of loss of KAT6A function. WM-8014 potentiates oncogene-induced senescence in vitro and in a zebrafish model of hepatocellular carcinoma. WM-1119, which has increased bioavailability, arrests the progression of lymphoma in mice. We anticipate that this class of inhibitors will help to accelerate the development of therapeutics that target gene transcription regulated by histone acetylation.
Sigma-Aldrich
WM-2474, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
WM-1119, ≥98% (HPLC)