MilliporeSigma
  • Cyst formation in proximal renal tubules caused by dysfunction of the microtubule minus-end regulator CAMSAP3.
Cyst formation in proximal renal tubules caused by dysfunction of the microtubule minus-end regulator CAMSAP3.
Scientific reports (2021-03-14)
Yuto Mitsuhata, Takaya Abe, Kazuyo Misaki, Yuna Nakajima, Keita Kiriya, Miwa Kawasaki, Hiroshi Kiyonari, Masatoshi Takeichi, Mika Toya, Masamitsu Sato
ABSTRACT
Epithelial cells organize an ordered array of non-centrosomal microtubules, the minus ends of which are regulated by CAMSAP3. The role of these microtubules in epithelial functions, however, is poorly understood. Here, we show that the kidneys of mice in which Camsap3 is mutated develop cysts at the proximal convoluted tubules (PCTs). PCTs were severely dilated in the mutant kidneys, and they also exhibited enhanced cell proliferation. In these PCTs, epithelial cells became flattened along with perturbation of microtubule arrays as well as of certain subcellular structures such as interdigitating basal processes. Furthermore, YAP and PIEZO1, which are known as mechanosensitive regulators for cell shaping and proliferation, were activated in these mutant PCT cells. These observations suggest that CAMSAP3-mediated microtubule networks are important for maintaining the proper mechanical properties of PCT cells, and its loss triggers cell deformation and proliferation via activation of mechanosensors, resulting in the dilation of PCTs.
Sigma-Aldrich
Anti-NF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-α-Tubulin−FITC antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Tubulin, Tyrosine antibody produced in mouse, clone TUB-1A2, ascites fluid
Sigma-Aldrich
Anti-α Galactosidase Antibody, Chemicon®, from chicken
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid