MilliporeSigma
  • Autoantibodies to amyloid beta-peptide (Abeta) are increased in Alzheimer's disease patients and Abeta antibodies can enhance Abeta neurotoxicity: implications for disease pathogenesis and vaccine development.
Autoantibodies to amyloid beta-peptide (Abeta) are increased in Alzheimer's disease patients and Abeta antibodies can enhance Abeta neurotoxicity: implications for disease pathogenesis and vaccine development.
Neuromolecular medicine (2003-04-01)
Avindra Nath, Elizabeth Hall, Marnia Tuzova, Michael Dobbs, Melina Jons, Caroline Anderson, Jerold Woodward, Zhihong Guo, Weiming Fu, Richard Kryscio, David Wekstein, Charles Smith, William R Markesbery, Mark P Mattson
ABSTRACT
Studies of amyloid precursor protein transgenic mice suggest that immune responses to amyloid beta peptide (Abeta) may be instrumental in the removal of plaques from the brain, but the initial clinical trial of an Abeta vaccine in patients with Alzheimer s disease (AD) was halted as the result of serious neurological complications in some patients. We now provide evidence that AD patients exhibit an enhanced immune response to Abeta and that, contrary to expectations, Abeta antibodies enhance the neurotoxic activity of the peptide. Serum titers to Abeta were significantly elevated in AD patients and Abeta antibodies were found in association with amyloid plaques in their brains, but there was no evidence of cell-mediated immune responses to Abeta in the patients. Abeta antibodies were detected in the serum of old APP mutant transgenic mice with plaque-like Abeta deposits, but not in the serum of younger transgenic or nontransgenic mice. Serum from APP mutant mice potentiated the neurotoxicity of Abeta. Our data suggest that a humoral immune response to Abeta in AD patients may promote neuronal degeneration, a process with important implications for the future of vaccine-based therapies for AD.
Sigma-Aldrich
Anti-Amyloid Antibody, β 1-16, clone DE2, culture supernatant, clone DE2, Chemicon®