MilliporeSigma
  • c-kit immunoreactive interstitial cells of Cajal in the human small and large intestine.
c-kit immunoreactive interstitial cells of Cajal in the human small and large intestine.
Histochemistry and cell biology (1998-04-16)
P Rømert, H B Mikkelsen
ABSTRACT
c-kit immunohistochemistry was performed on unfixed frozen sections of human small (duodenum, jejunum, and ileum) and large intestine (ascending, transverse, descending, and sigmoid colon). The c-kit immunoreactive cells in the muscularis externa of the intestinal wall were identified as interstitial cells of Cajal (ICC) and mast cells. ICC were identified by their morphology, localization, and organization based on previous light and electron microscopic studies. In the small intestine, ICC were located primarily in relation to the myenteric plexus of Auerbach, but also in septa between circular muscle lamellae. In the large intestine, ICC were seen in relation to Auerbach's plexus, but also and in great numbers in the circular muscle layer and in teniae of the longitudinal muscle layer. The morphology of the ICC was similar in the small and large intestine, but the pattern of distribution was obviously different. c-kit immunoreactive mast cells were found predominantly in the inner part of the circular muscle layer. The anti-c-kit method is found to be an easy and reliable method to study at least most of the interstitial cells of Cajal and thereby contribute to further normal and pathological studies.
Sigma-Aldrich
Anti-Tryptase Antibody, Mast Cell, clone G3, clone G3, Chemicon®, from mouse