MilliporeSigma
  • Evaluation of Tegaran Formula ZhenHua cytotoxicity against human cancer cell lines.
Evaluation of Tegaran Formula ZhenHua cytotoxicity against human cancer cell lines.
PloS one (2020-10-22)
Panagiotis Parsonidis, Ioanna Vlachou, Alexandra Mamagkaki, Ioannis Bouris, Vasiliki Daikopoulou, Ioannis Papasotiriou
ABSTRACT
The aim of this study is to evaluate the potential health effects of Tegaran Formula ZhenHua, a nutritional supplement used mainly by cancer patients. Its active ingredients and cytotoxicity was assessed with analytical methods and viability assays, respectively. The analytical methods consisted of dissolution, disintegration, HPLC, LC/MS, GC/MS and NMR. Cytotoxicity was assessed by MTT, SRB, CVE colorimetric viability assays in 0, 24, 48 and 72h time points. The results indicate that Tegaran Formula ZhenHua supplement did not present any cytotoxic effects due to issues related to the capsules' solubility, distribution and identification of the active ingredient.
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Supelco
Daidzein, analytical standard
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Sulforhodamine B, acid form, laser grade, Dye content 95 %
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
COLO 699 N