MilliporeSigma
  • Synergistic activity of R207910 combined with pyrazinamide against murine tuberculosis.
Synergistic activity of R207910 combined with pyrazinamide against murine tuberculosis.
Antimicrobial agents and chemotherapy (2006-12-21)
M Ibrahim, K Andries, N Lounis, A Chauffour, C Truffot-Pernot, V Jarlier, N Veziris
ABSTRACT
In previous studies, the diarylquinoline R207910 (also known as TMC207) was demonstrated to have high bactericidal activity when combined with first- or second-line antituberculous drugs. Here we extend the evaluation of R207910 in the curative model of murine tuberculosis by assessing the activities of one-, two-, and three-drug combinations containing R207910 and isoniazid (INH), rifampin (RIF), pyrazinamide (PZA), or moxifloxacin (MXF) in the setting of a high initial bacillary load (7.2 log(10) CFU). Two months of treatment with the combinations R207910-PZA, R207910-PZA-INH, R207910-PZA-RIF, or R207910-PZA-MXF resulted in culture-negative lung homogenates in 70 to 100% of the mice, while mice treated with INH-RIF-PZA (the reference regimen) or RIF-MXF-PZA remained culture positive. Combinations including R207910 but not PZA (e.g., R207910-INH-RIF and R207910-MXF-RIF) were less active than R207910-PZA-containing regimens administered either alone or with the addition of INH, RIF, or MXF. These results reveal a synergistic interaction between R207910 and PZA. Three-drug combinations containing these two drugs and INH, RIF, or MXF have the potential to significantly shorten the treatment duration in patients, provided that these results can be confirmed in long-term experiments including periods of relapse.
Sigma-Aldrich
TMC207, ≥98% (HPLC)
Sigma-Aldrich
Pyrazinecarboxamide
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Supelco
Isoniazid, analytical standard, ≥99% (TLC)