MilliporeSigma
  • Dopaminergic Control of Striatal Cholinergic Interneurons Underlies Cocaine-Induced Psychostimulation.
Dopaminergic Control of Striatal Cholinergic Interneurons Underlies Cocaine-Induced Psychostimulation.
Cell reports (2020-04-23)
Robert G Lewis, Marcello Serra, Daniela Radl, Michela Gori, Catherine Tran, Sharon E Michalak, Christopher D Vanderwal, Emiliana Borrelli
ABSTRACT
Cocaine drastically elevates dopamine (DA) levels in the striatum, a brain region that is critical to the psychomotor and rewarding properties of the drug. DA signaling regulates intrastriatal circuits connecting medium spiny neurons (MSNs) with afferent fibers and interneurons. While the cocaine-mediated increase in DA signaling on MSNs is well documented, that on cholinergic interneurons (ChIs) has been more difficult to assess. Using combined pharmacological, chemogenetic, and cell-specific ablation approaches, we reveal that the D2R-dependent inhibition of acetylcholine (ACh) signaling is fundamental to cocaine-induced changes in behavior and the striatal genomic response. We show that the D2R-dependent control of striatal ChIs enables the motor, sensitized, and reinforcing properties of cocaine. This study highlights the importance of the DA- and D2R-mediated inhibitory control of ChIs activity in the normal functioning of striatal networks.
Sigma-Aldrich
Donepezil hydrochloride, ≥98% (HPLC)
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Roche
Fluorescein RNA Labeling Mix, solution, suitable for Northern blotting, suitable for Southern blotting, suitable for hybridization
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Roche
Anti-Fluorescein-POD, Fab fragments, from sheep