MilliporeSigma
  • HP1γ regulates H3K36 methylation and pluripotency in embryonic stem cells.
HP1γ regulates H3K36 methylation and pluripotency in embryonic stem cells.
Nucleic acids research (2020-11-26)
Nur Zafirah Zaidan, Rupa Sridharan
ABSTRACT
The heterochromatin protein 1 (HP1) family members are canonical effectors and propagators of gene repression mediated by histone H3 lysine 9 (H3K9) methylation. HP1γ exhibits an increased interaction with active transcription elongation-associated factors in embryonic stem cells (ESCs) compared to somatic cells. However, whether this association has a functional consequence remains elusive. Here we find that genic HP1γ colocalizes and enhances enrichment of transcription elongation-associated H3K36me3 rather than H3K9me3. Unexpectedly, sustained H3K36me3 deposition is dependent on HP1γ. HP1γ-deleted ESCs display reduced H3K36me3 enrichment, concomitant with decreased expression at shared genes which function to maintain cellular homeostasis. Both the H3K9me3-binding chromodomain and histone binding ability of HP1γ are dispensable for maintaining H3K36me3 levels. Instead, the chromoshadow together with the hinge domain of HP1γ that confer protein and nucleic acid-binding ability are sufficient because they retain the ability to interact with NSD1, an H3K36 methyltransferase. HP1γ-deleted ESCs have a slower self-renewal rate and an impaired ability to differentiate towards cardiac mesoderm. Our findings reveal a requirement for HP1γ in faithful establishment of transcription elongation in ESCs, which regulates pluripotency.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-NSD1 Antibody, clone 1NW-1A10, ascites fluid, clone 1NW-1A10, from mouse