MilliporeSigma
  • Form of calf diet and the rumen. II: Impact on volatile fatty acid absorption.
Form of calf diet and the rumen. II: Impact on volatile fatty acid absorption.
Journal of dairy science (2019-07-08)
T T Yohe, H Schramm, R R White, M D Hanigan, C L M Parsons, H L M Tucker, B D Enger, N R Hardy, K M Daniels
ABSTRACT
Diet is known to affect rumen growth and development. Calves fed an all-liquid diet have smaller and less developed rumens and a decreased ability to absorb volatile fatty acids (VFA) compared to calves fed both liquid and dry feed. However, it is unknown how rumens respond when challenged with a defined concentration of VFA. The objective of this study was to assess the effects of 2 different feeding programs on VFA absorption in preweaned calves. Neonatal Holstein bull calves were individually housed and randomly assigned to 1 of 2 diets. The diets were milk replacer only (MRO; n = 5) or milk replacer with starter (MRS; n = 6). Diets were isoenergetic (3.87 ± 0.06 Mcal of metabolizable energy per day) and isonitrogenous (0.17 ± 0.003 kg/d of apparent digestible protein). Milk replacer was 22% crude protein, 21.5% fat (dry matter basis). The textured calf starter was 21.5% crude protein (dry matter basis). Feed and ad libitum water intakes were recorded daily. Calves were exposed to a defined concentration of VFA buffer (acetate 143 mM, propionate 100 mM, butyrate 40.5 mM) 6 h before euthanasia on d 43 ± 1. Rumen fluid samples were obtained every 15 to 30 min for 6 h to measure the rate of VFA absorption. Rumen tissues were obtained from the ventral sac region and processed for morphological and immunohistochemical analyses of the VFA transporters monocarboxylate transporter 1 (MCT1) and 4 (MCT4). Body growth did not differ between diets, but empty reticulorumens were heavier in MRS than MRO calves (0.67 vs. 0.39 ± 0.04 kg) and MRS calves had larger papillae areas (0.76 vs. 15 ± 0.08 mm2). We observed no differences between diets in terms of the abundance of MCT1 and MCT4 per unit area. These results indicate that the extrapolated increase in total abundance of MCT1 or MCT4 in MRS calves was not due to increased transporter density per unit area. Modeled VFA absorption metrics (flux, mmol/h, or 6 h absorbed VFA in mmol) were not different across diets. These results demonstrate that the form of calfhood diet, whether solely MR or MR and starter, does not alter VFA absorption capacity when the rumen is exposed to a defined concentration of VFA at 6 wk of age.
Sigma-Aldrich
Anti-Monocarboxylate Transporter 1 Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-Monocarboxylate Transporter 4 Antibody, Chemicon®, from rabbit