MilliporeSigma
  • Microtubule-dependent processes precede pathological calcium influx in excitotoxin-induced axon degeneration.
Microtubule-dependent processes precede pathological calcium influx in excitotoxin-induced axon degeneration.
Journal of neurochemistry (2019-11-11)
Nan Tian, Kelsey A Hanson, Alison J Canty, James C Vickers, Anna E King
ABSTRACT
Axon degeneration and axonal loss is a feature of neurodegenerative disease and injury and occurs via programmed pathways that are distinct from cell death pathways. While the pathways of axonal loss following axon severing are well described, less is known about axonal loss following other neurodegenerative insults. Here we use primary mouse cortical neuron cultures grown in compartmentalized chambers to investigate the role of calcium in the degeneration of axons that occurs following a somal insult by the excitotoxin kainic acid. Calcium influx has been implicated in both excitotoxicity and axon degeneration mechanisms, however the link between a somal insult and axonal calcium increase is unclear. Live imaging of axons demonstrated that pharmacologically preventing intracellular calcium increases through the endoplasmic reticulum or mitochondria significantly (p < 0.05) reduced axon degeneration. Live calcium-imaging with the Ca2+ indicator Fluo-4 demonstrated that kainic acid exposure to the soma resulted in a rapid, and transient, increase in calcium in the axon, which occured even at low kainic acid concentrations that do not cause axon degeneration within 24 h. However, this calcium transient was followed by a gradual increase in axonal calcium, which was associated with axonal loss. Furthermore, treatment with a range of doses of the microtubule stabilizing drug taxol, which protects against axon fragmentation in this model, prevented this gradual calcium increase, suggesting that the intra-axonal calcium changes are downstream of microtubule associated events. Biochemical analysis of taxol treated neurons demonstrated a shift in microtubule post-translational modifications, with a significant (p < 0.05) increase in acetylated tubulin and a significant (p < 0.05) decrease in tyrosinated tubulin, suggestive of a more stable microtubule pool. Together our results suggest that axonal degeneration following excitotoxicity is dependent on an increase in axonal calcium, which is downstream of a microtubule-dependent event.
Sigma-Aldrich
Kainic acid monohydrate, ≥99% (TLC)
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Dantrolene sodium salt
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Anti-alpha Tubulin Antibody, tyrosinated, serum, from rabbit
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)