MilliporeSigma
The deubiquitinase JOSD2 is a positive regulator of glucose metabolism.
Cell death and differentiation (2020-10-22)
Lyudmila Krassikova, Boxi Zhang, Divya Nagarajan, André Lima Queiroz, Merve Kacal, Evangelos Samakidis, Helin Vakifahmetoglu-Norberg, Erik Norberg, Lyudmila Krassikova, Boxi Zhang, Divya Nagarajan, André Lima Queiroz, Merve Kacal, Evangelos Samakidis, Helin Vakifahmetoglu-Norberg, Erik Norberg
ABSTRACT
Cancer cells undergo complex metabolic alterations. The mechanisms underlying the tuning of cancer metabolism are under active investigation. Here, we identify the uncharacterized deubiquitinase JOSD2 as a positive regulator of cancer cell proliferation by displaying comprehensive effects on glucose catabolism. We found that JOSD2 directly controls a metabolic enzyme complex that includes Aldolase A, Phosphofructokinase-1 and Phosphoglycerate dehydrogenase, in vitro and in vivo. Further, JOSD2 expression, but not a catalytically inactive mutant, deubiquitinates and stabilizes the enzyme complex, thereby enhancing their activities and the glycolytic rate. This represents a selective JOSD2 feature that is not shared among other Machado-Joseph disease DUBs or observed in nontransformed cells. JOSD2 deficiency displays cytostatic effects and reduces glycolysis in a broad spectrum of tumor cells of distinct origin and its expression correlates with poor prognosis in non-small cell lung cancer. Overall, our study provides evidence for a previously unknown biological mechanism in which JOSD2 integrates glucose and serine metabolism with potential therapeutic implications.
Sigma-Aldrich
Phosphofructokinase (PFK) Activity Colorimetric Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
Anti-PFKM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PHGDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1