MilliporeSigma
  • Thermoresponsive polysaccharides with tunable thermoresponsive properties via functionalisation with alkylamide groups.
Thermoresponsive polysaccharides with tunable thermoresponsive properties via functionalisation with alkylamide groups.
Carbohydrate polymers (2020-12-29)
Sarah Otto, Paula Facal Marina, Fiona Zhou, Anton Blencowe
ABSTRACT
Polysaccharides have been used widely in many industries, from food technology and mining to cosmetics and biomedical applications. Over recent years there has been growing interest in the development of responsive polysaccharides with unique and switchable properties, particularly systems that display lower-critical solution temperatures (LCSTs). Therefore, in this study we aimed to investigate a novel strategy that would allow the conversion of non-responsive polysaccharides into thermoresponsive polysaccharides with tuneable LCSTs. Through the functionalisation of dextran with alkylamide groups (isopropyl amide, diethyl amide, piperidinyl and diisobutyl amide) using a carbodiimide coupling approach in conjunction with amic acid derivatives, we prepared a library of novel dextrans with various degrees of substitution (DS), which were characterised via nuclear magnetic resonance (NMR) spectroscopy and gel permeation chromatography (GPC). The alkylamide-functionalised dextrans were found to have good solubility in aqueous solutions, with the exception of those having a high DS of large hydrophobic substituents. Determination of the thermoresponsive characteristics of the polymer solutions via UV-vis spectroscopy revealed that the LCST of the alkylamide-functionalised dextrans was highly dependent on the type of alkylamide group and the DS and could be tuned over a large range (5-35 °C). Above the LCST, all of the thermoresponsive alkylamide-functionalised dextrans formed colloidal dispersions with particles sizes ranging from 400 -600 nm, as determined by dynamic light scattering (DLS). In addition, the polymers were found to exhibit a fast and reversible phase transition in solution with narrow hysteresis (∼ 1-5 °C). Finally, the injectability and biocompatibility of the novel thermoresponsive dextrans was confirmed in vivo via subcutaneous and intracranial ventricle injections, with no local or systemic toxicity noted over a 14 d period. Overall, the alkylamide-functionalised dextrans display interesting thermoresponsive properties and trends that may make them useful in biomedical applications, such as drug-delivery.
Sigma-Aldrich
Diisobutylamine, 99%
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, histological tissue fixative