MilliporeSigma
  • Sequential purification and characterization of Torpedo californica nAChR-DC supplemented with CHS for high-resolution crystallization studies.
Sequential purification and characterization of Torpedo californica nAChR-DC supplemented with CHS for high-resolution crystallization studies.
Analytical biochemistry (2020-08-09)
Rafael Maldonado-Hernández, Orestes Quesada, José O Colón-Sáez, José A Lasalde-Dominicci, Rafael Maldonado-Hernández, Orestes Quesada, José O Colón-Sáez, José A Lasalde-Dominicci
ABSTRACT
Over the past 10 years we have been developing a multi-attribute analytical platform that allows for the preparation of milligram amounts of functional, high-pure, and stable Torpedo (muscle-type) nAChR detergent complexes for crystallization purpose. In the present work, we have been able to significantly improve and optimize the purity and yield of nicotinic acetylcholine receptors in detergent complexes (nAChR-DC) without compromising stability and functionality. We implemented new methods in the process, such as analysis and rapid production of samples for future crystallization preparations. Native nAChR was extracted from the electric organ of Torpedo californica using the lipid-like detergent LysoFos Choline 16 (LFC-16), followed by three consecutive steps of chromatography purification. We evaluated the effect of cholesteryl hemisuccinate (CHS) supplementation during the affinity purification steps of nAChR-LFC-16 in terms of receptor secondary structure, stability and functionality. CHS produced significant changes in the degree of β-secondary structure, these changes compromise the diffusion of the nAChR-LFC-16 in lipid cubic phase. The behavior was reversed by Methyl-β-Cyclodextrin treatment. Also, CHS decreased acetylcholine evoked currents of Xenopus leavis oocyte injected with nAChR-LFC-16 in a concentration-dependent manner. Methyl-β-Cyclodextrin treatment do not reverse functionality, however column delipidation produced a functional protein similar to nAChR-LFC-16 without CHS treatment.
Sigma-Aldrich
Cholesteryl hemisuccinate
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Millipore
Methyl α-D-mannopyranoside, ≥99.0%, suitable for microbiology, enables differentiation between species of Listeria
Sigma-Aldrich
Bromoacetylcholine bromide