MilliporeSigma
  • Immune response of horses to inactivated African horse sickness vaccines.
Immune response of horses to inactivated African horse sickness vaccines.
BMC veterinary research (2020-09-03)
Marina Rodríguez, Sunitha Joseph, Martin Pfeffer, Rekha Raghavan, Ulrich Wernery
ABSTRACT
African horse sickness (AHS) is a serious viral disease of equids resulting in the deaths of many equids in sub-Saharan Africa that has been recognized for centuries. This has significant economic impact on the horse industry, despite the good husbandry practices. Currently, prevention and control of the disease is based on administration of live attenuated vaccines and control of the arthropod vectors. A total of 29 horses in 2 groups, were vaccinated. Eighteen horses in Group 1 were further divided into 9 subgroups of 2 horses each, were individually immunised with one of 1 to 9 AHS serotypes, respectively. The eleven horses of Group 2 were immunised with all 9 serotypes simultaneously with 2 different vaccinations containing 5 serotypes (1, 4, 7-9) and 4 serotypes (2, 3, 5, 6) respectively. The duration of this study was 12 months. Blood samples were periodically withdrawn for serum antibody tests using ELISA and VNT and for 2 weeks after each vaccination for PCR and virus isolation. After the booster vaccination, these 27 horses seroconverted, however 2 horses responded poorly as measured by ELISA. In Group 1 ELISA and VN antibodies declined between 5 to 7 months post vaccination (pv). Twelve months later, the antibody levels in most of the horses decreased to the seronegative range until the annual booster where all horses again seroconverted strongly. In Group 2, ELISA antibodies were positive after the first booster and VN antibodies started to appear for some serotypes after primary vaccination. After booster vaccination, VN antibodies increased in a different pattern for each serotype. Antibodies remained high for 12 months and increased strongly after the annual booster in 78% of the horses. PCR and virus isolation results remained negative. Horses vaccinated with single serotypes need a booster after 6 months and simultaneously immunised horses after 12 months. Due to the non-availability of a facility in the UAE, no challenge infection could be carried out.
Sigma-Aldrich
2-Bromoethylamine hydrobromide, 99%