MilliporeSigma
  • Immunotherapy for nonalcoholic steatohepatitis using the multiple cytokine production modulator Y-40138.
Immunotherapy for nonalcoholic steatohepatitis using the multiple cytokine production modulator Y-40138.
World journal of gastroenterology (2009-11-26)
Tatsuhiro Tsujimoto, Hideto Kawaratani, Toshiyuki Kitazawa, Hitoshi Yoshiji, Masao Fujimoto, Masahito Uemura, Hiroshi Fukui
ABSTRACT
To investigate the possible use of the multiple cytokine production modulator, Y-40138, as a novel immunotherapy in the rat nonalcoholic steatohepatitis (NASH) model. We allocated 6-wk-old male F344 rats to choline-supplemented, L-amino acid-defined (CSAA) diet (control group), CSAA diet + Y-40138 (control + Y-40138 group), choline-deficient, L-amino acid-defined (CDAA) diet (NASH group), or CDAA diet + Y-40138 (NASH + Y-40138 group). In each group, we measured the plasma alanine aminotransferase (ALT) levels, and the plasma and liver levels of tumor necrosis factor-alpha (TNF-alpha), interferon-gamma (IFN-gamma), and interleukin-10 (IL-10). Tissue specimens of phosphate buffered saline-perfused liver were subjected to hematoxylin and eosin staining, Azan staining, Sirius red staining, and immunohistochemical staining (for Kupffer cells and TNF-alpha). We then extracted Kupffer cells from the collagenase-perfused livers using the Percoll gradient centrifugation method, and measured the TNF-alpha levels in the supernatant (in vitro TNF-alpha production by Kupffer cells) using an enzyme-linked immunosorbent assay kit. In comparison to the NASH group, serum ALT elevation was mild, production of serum and liver TNF-alpha and IFN-gamma was inhibited, and IL-10 production was increased in the NASH + Y-40138 group. Amelioration of liver histology was also noted in the NASH + Y-40138 group. Kupffer cell immunohistochemical staining revealed no differences between groups, whereas TNF-alpha immunohistochemical staining showed fewer stained cells in the NASH + Y-40138 group than in the NASH group. The TNF-alpha levels in the in-vitro Kupffer cell culture supernatant were lower in the NASH + Y-40138 group than in the NASH group. Administration of Y-40138 to NASH model rats reduced hepatic inflammation and suppressed fibrosis. These results indicate that the multiple cytokine production modulator, Y-40138, is promising as a novel treatment for NASH.
Product Description
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s salts, with 2.0 mM L-glutamine, with 20.0 mM HEPES, with non-essential amino acids, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with non-essential amino acids, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 25mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, with non-essential amino acids, without sodium bicarbonate, dry powder, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, without L-glutamine, liquid, sterile-filtered, suitable for cell culture