MilliporeSigma
  • SYNTHESIS OF 1,3,4-OXADIAZOLES AS SELECTIVE T-TYPE CALCIUM CHANNEL INHIBITORS.
SYNTHESIS OF 1,3,4-OXADIAZOLES AS SELECTIVE T-TYPE CALCIUM CHANNEL INHIBITORS.
Heterocycles (2020-08-11)
Man Zhang, Bende Zou, Medha J Gunaratna, Sahani Weerasekara, Zongbo Tong, Thi D T Nguyen, Serkan Koldas, William S Cao, Conrado Pascual, Xinmin Simon Xie, Duy H Hua
ABSTRACT
Neuropathic pain, epilepsy, insomnia, and tremor disorder may arrive from an increase of intracellular Ca2+ concentration through a dysfunction of T-type Ca2+ channels. Thus, T-type calcium channels could be a target in drug discovery for the treatments of neuropathic pain and epilepsy. From rational drug design approach, a group of 2,5-disubstituted 1,3,4-oxadiazole molecules was synthesized and their selective T-type channel inhibitions were evaluated. The synthetic strategy consists of a short sequence of three reactions: (i) condensation of thiosemicarbazide with acid chlorides; (ii) ring closing by 1,3-dibromo-5,5- dimethylhydantoin; and (iii) coupling with various acid chlorides. 5-Chloro-N-(5- phenyl-1,3,4-oxadiazol-2-yl)thiophene-2-carboxamide (11) was found to selectively inhibit T-type Ca2+ channel over Na+ and K+ channels in mouse dorsal root ganglion neurons and/or human embryonic kidney (HEK)-293 cells and to suppress seizure-induced death in mouse model. Consequently, compound 11 is a useful probe for investigation of physiologic and pathophysiologic roles of the T-channel, and provides a basis to develop a novel therapeutic to treat chronic neuropathic and inflammatory pains.
Sigma-Aldrich
5-Chlorothiophene-2-carboxylic acid, 97%