MilliporeSigma
The mechanisms and clinical significance of PDCD4 in colorectal cancer.
Gene (2018-09-24)
Jiali Long, Yuting Yin, Haina Guo, Shuling Li, Yanqin Sun, Chao Zeng, Wei Zhu
ABSTRACT
In recent years, the incidence and mortality of colorectal cancer (CRC) have been on a global upward trend. There is an urgent need for effective tools to prevent and treat CRC and reduce morbidity and mortality of CRC patients. Recent evidence suggests that programmed cell death 4 (PDCD4), a novel tumor suppressor gene, inhibits tumor progression at transcriptional and translational levels and regulates multiple signal transduction pathways. However, little is known about the precise mechanisms regulating PDCD4 expression in CRC. In addition, several studies have demonstrated that the expression of in CRC is down-regulated or even absent. PDCD4 is therefore considered to be an independent prognostic factor in CRC and may be a potential support diagnostic tool for distinguishing in normal colon tissue, benign adenoma and CRC. This review will focus on the expression of PDCD4 in CRC and the relevant molecular mechanisms.