MilliporeSigma
  • Quantitative analysis of oxytetracycline and its 4-epimer in calf tissues by high-performance liquid chromatography combined with positive electrospray ionization mass spectrometry.
Quantitative analysis of oxytetracycline and its 4-epimer in calf tissues by high-performance liquid chromatography combined with positive electrospray ionization mass spectrometry.
The Analyst (2003-08-05)
Marc Cherlet, Siegrid De Baere, Patrick De Backer
ABSTRACT
Tetracycline antibiotics are commonly used in veterinary medicine because of their broad spectrum activity and cost effectiveness. Oxytetracycline (OTC) is one of the most important members of this antibiotic family. The purpose of this study was to develop and validate a method to determine OTC residues in edible tissues of calf. Extraction of OTC and its 4-epimer (4-epiOTC), in the presence of the internal standard demethylchlortetracycline (DMCTC), was performed using a liquid extraction with sodium succinate solution (pH 4.0), followed by protein removal with trichloroacetic acid and paper filtration. Further solid-phase extraction clean-up on an HLB polymeric reversed phase column was performed to obtain an extract suitable for LC-MS-MS analysis. Chromatographic separation of the internal standard, and especially OTC and its 4-epimer, was achieved on a PLRP-S polymeric reversed phase column, using a mixture of 0.001 M of oxalic acid, 0.5% (v/v) of formic acid and 3% (v/v) of tetrahydrofuran in water (mobile phase A) and tetrahydrofuran (mobile phase B) as the mobile phase, and at a column temperature of 60 degrees C. OTC and its 4-epimer could be identified using the MS-MS detection technique, and were subsequently quantified. The method has been validated according to the requirements of the EC at the MRL (maximum residue limit, 100 ng g(-1) for muscle, 300 ng g(-1) for liver and600 ng g(-1) for kidney), half the MRL and double the MRL levels, as well for OTC as for 4-epiOTC. Calibration graphs were prepared for all tissues and good linearity was achieved over the concentration ranges tested (r > 0.99 and goodness of fit < 10%). Limits of quantification of half the MRLs were obtained for the analysis of OTC and 4-epiOTC in muscle, liver and kidney tissues of calf. Limits of detection ranged for both components between 0.8 and 48.2 ng g(-1). The within-day and between-day precisions, expressed as RSD values, were all below the maximum allowed RSD values calculated according to the Horwitz equation. The results for accuracy fell within the -20% to +10% range. Recoveries were between 47 and 56% for OTC, and between 52 and 62% for 4-epiOTC, depending on the tissue. The method has been successfully used for the quantitative determination of OTC and 4-epiOTC in tissue samples of calves medicated with OTC by intramuscular injection.
Sigma-Aldrich
Oxytetracycline dihydrate, Assay (anhydrous basis) 95.0%-102.0%