MilliporeSigma
  • Continuous-flow hydrogenation of carbon dioxide to pure formic acid using an integrated scCO2 process with immobilized catalyst and base.
Continuous-flow hydrogenation of carbon dioxide to pure formic acid using an integrated scCO2 process with immobilized catalyst and base.
Angewandte Chemie (International ed. in English) (2012-07-19)
Sebastian Wesselbaum, Ulrich Hintermair, Walter Leitner
ABSTRACT
Dual role for CO(2): Pure formic acid can be obtained continuously by hydrogenation of CO(2) in a single processing unit. An immobilized ruthenium organometallic catalyst and a nonvolatile base in an ionic liquid (IL) are combined with supercritical CO(2) as both reactant and extractive phase.
Sigma-Aldrich
QuadraPure® TU, macroporous, 400-600 μm particle size