MilliporeSigma
  • Live Visualization of Hemagglutinin Dynamics During Infection by Using a Novel Reporter Influenza A Virus.
Live Visualization of Hemagglutinin Dynamics During Infection by Using a Novel Reporter Influenza A Virus.
Viruses (2020-07-02)
Luiz Gustavo Dos Anjos Borges, Giuseppe Pisanelli, Oyahida Khatun, Adolfo García-Sastre, Shashank Tripathi
ABSTRACT
Live visualization of influenza A virus (IAV) structural proteins during viral infection in cells is highly sought objective to study different aspects of the viral replication cycle. To achieve this, we engineered an IAV to express a Tetra Cysteine tag (TC tag) from hemagglutinin (HA), which allows intracellular labeling of the engineered HA protein with biarsenic dyes and subsequent fluorescence detection. Using such constructs, we rescued a recombinant IAV with TC tag inserted in HA, in A/Puerto Rico/8/1934(H1N1) background (HA-TC). This recombinant HA-TC tag reporter IAV was replication-competent; however, as compared to wild type PR8 IAV, it was attenuated in multicycle replication. We confirmed expression of TC tag and biarsenical labeling of HA by immunofluorescence assay in cells infected with an HA-TC tag reporter IAV. Further, we used this reporter virus to visualize HA expression and translocation in IAV infected cells by live confocal imaging. We also tested the utility of the HA-TC IAV in testing chemical inhibitors of the HA translocation. Overall, HA-TC IAV is a versatile tool that will be useful for studying viral life cycle events, virus-host interactions, and anti-viral testing.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Supelco
Perfluoroundecanoic acid, analytical standard