MilliporeSigma
  • Morphological Transitions of Photoresponsive Vesicles from Amphiphilic Polypeptoid Copolymers for Controlled Release.
Morphological Transitions of Photoresponsive Vesicles from Amphiphilic Polypeptoid Copolymers for Controlled Release.
Polymers (2020-04-09)
Xu Yang, Zhiwei Wang, Jing Sun
ABSTRACT
Photoresponsive polymers have attracted increasing interest for a variety of applications. Here, we report a family of photoresponsive polypeptoid-based copolymer poly(ethylene glycol)-b-poly(N-(S-(o-nitrobenzyl)-thioethyl) glycine)-co-poly(N-(2-phenylethyl) glycine) (PEG-b-PNSN-co-PNPE) synthesized by the controlled ring-opening polymerization (ROP) technique. The key feature of the design is to incorporate both o-nitrobenzyl group moiety to offer the photoresponsive property and phenethyl residues to tune the structural and amphiphilic property of the system. We demonstrate that the cleavage degree of the o-nitrobenzyl group can reach to 100% upon UV-irradiation. With delicate design, a photoresponsive vesicle-to-sphere transition has been observed that facilitates the release of the encapsulants. This work provides a facile approach to prepare a type of photoresponsive polymers with tunable properties for drug delivery.
Sigma-Aldrich
2-Nitrobenzyl bromide, 98%