MilliporeSigma
Oxyresveratrol Possesses DNA Damaging Activity.
Molecules (Basel, Switzerland) (2020-06-05)
Sarayut Radapong, Satyajit D Sarker, Kenneth J Ritchie
ABSTRACT
Artocarpus lakoocha Wall. ex Roxb. (family: Moraceae) has been used as a traditional Thai medicine for the treatment of various parasitic diseases. This species has been reported to be the source of phytochemicals, which show potent biological activities. The objective of this study was to investigate the phytochemical profile of the extracts of the heartwood of A. lakoocha and their pro-oxidant activity in vitro. The heartwood was ground, extracted, and then chromatographic and spectroscopic analyses were carried out; oxyresveratrol was identified as the major component in the extracts. The pro-oxidant activity was investigated using DNA-nick, reactive oxygen species and reducing assays. The results showed that oxyresveratrol induced DNA damage dose-dependently in the presence of copper (II) ions. It was also found to generate reactive oxygen species (ROS) in a dose-dependent manner and reduce copper (II) to copper (I). It is concluded that oxyresveratrol is the most abundant stilbenoid in A. lakoocha heartwood. The compound exhibited pro-oxidant activity in the presence of copper (II) ions, which may be associated with its ability to act as an anticancer compound.
Sigma-Aldrich
Oxyresveratrol, ≥97.0% (HPLC)
Sigma-Aldrich
Copper(II) acetate, powder, 99.99% trace metals basis