MilliporeSigma
  • Simultaneous Control of Endogenous and User-Defined Genetic Pathways Using Unique ecDHFR Pharmacological Chaperones.
Simultaneous Control of Endogenous and User-Defined Genetic Pathways Using Unique ecDHFR Pharmacological Chaperones.
Cell chemical biology (2020-04-25)
Prerana Ramadurgum, DaNae R Woodard, Steffi Daniel, Hui Peng, Prema L Mallipeddi, Hanspeter Niederstrasser, Melina Mihelakis, Viet Q Chau, Peter M Douglas, Bruce A Posner, John D Hulleman
ABSTRACT
Destabilizing domains (DDs), such as a mutated form of Escherichia coli dihydrofolate reductase (ecDHFR), confer instability and promote protein degradation. However, when combined with small-molecule stabilizers (e.g., the antibiotic trimethoprim), DDs allow positive regulation of fusion protein abundance. Using a combinatorial screening approach, we identified and validated 17 unique 2,4-diaminopyrimidine/triazine-based ecDHFR DD stabilizers, at least 15 of which were ineffective antibiotics against E. coli and S. aureus. Identified stabilizers functioned in vivo to control an ecDHFR DD-firefly luciferase in the mouse eye and/or the liver. Next, stabilizers were leveraged to perform synergistic dual functions in vitro (HeLa cell death sensitization) and in vivo (repression of ocular inflammation) by stabilizing a user-defined ecDHFR DD while also controlling endogenous signaling pathways. Thus, these newly identified pharmacological chaperones allow for simultaneous control of compound-specific endogenous and user-defined genetic pathways, the combination of which may provide synergistic effects in complex biological scenarios.
Sigma-Aldrich
2-Amino-3H-quinazolin-4-one, AldrichCPR
Sigma-Aldrich
2,4-Diaminoquinazoline, AldrichCPR
Sigma-Aldrich
Dihydrofolic acid, ≥90%
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable (for use in cellulose acetate electrophoresis), 0.1 % (w/v) in 5% acetic acid
Magnetic Stir Bar, for use with For 3.0-5.0mL vessels, pkg of 6 ea, PTFE
Sigma-Aldrich
Dihydrofolate Reductase Assay Kit, 1 kit sufficient for 50-100 tests
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture