MilliporeSigma
  • Induction of ASC pyroptosis requires gasdermin D or caspase-1/11-dependent mediators and IFNβ from pyroptotic macrophages.
Induction of ASC pyroptosis requires gasdermin D or caspase-1/11-dependent mediators and IFNβ from pyroptotic macrophages.
Cell death & disease (2020-06-20)
Cuiping Zhang, Caiqi Zhao, Xiaoyan Chen, Rujia Tao, Sijiao Wang, Guangxun Meng, Xing Liu, Changzhou Shao, Xiao Su
ABSTRACT
Mesenchymal stem cells (MSCs) have been used in cell-based therapies for a variety of disorders. Some factors such as inflammatory mediators in the diseased area might damage the survival of MSCs and affect their efficacy. Pyroptosis is a form of programmed necrosis as a response for immune cells to cytosolic pathogenic stimuli. Whether MSCs develop pyroptosis under pathological stimulation, its underlying mechanism and biological significance are still unclear. Here, we found that LPS, flagellin, dsDNA, nigericin (NIG), or LPS combined with nigericin (LPS/NIG) could not induce pyroptosis in adipose-tissue-derived mesenchymal stem cells (ASCs). However, when applied the culture media collected from LPS/NIG-induced pyroptotic bone marrow-derived macrophages (BMDMs) to incubate ASCs, ASCs developed pyroptosis. Inhibition of caspases or deletion of Caspase-1/11 in ASCs did not affect the pyroptotic macrophage media-triggered ASC pyroptosis while ablation of Caspase-1/11 abolished BMDM pyroptosis induced by LPS/NIG. Media collected from LPS/NIG stimulated Gsdmd-/- or Caspase-1/11-/- BMDMs could not induce pyroptosis of ASCs. In addition, RNA-seq analysis showed that interferon (IFN)-stimulated genes were upregulated in pyroptotic ASCs. Adding IFNβ could boost LPS/NIG stimulated BMDM media-induced ASC pyroptosis. Surprisingly, the pyroptotic ASCs had a lower bactericidal ability to P. Aeruginosa. Taken together, induction of ASC pyroptosis requires gasdermin D or caspase-1/11-dependent mediators and IFNβ from pyroptotic macrophages.
Sigma-Aldrich
Lipopolysaccharides from Pseudomonas aeruginosa 10, purified by phenol extraction
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid