MilliporeSigma
  • Degradable pH-responsive NIR-II imaging probes based on a polymer-lanthanide composite for chemotherapy.
Degradable pH-responsive NIR-II imaging probes based on a polymer-lanthanide composite for chemotherapy.
Dalton transactions (Cambridge, England : 2003) (2020-07-01)
Miao Feng, Yanxing Wang, Bi Lin, Xiangrong Peng, Ying Yuan, Xiaofeng Tao, Ruichan Lv
ABSTRACT
In this research, a pH-sensitive degradable nanoprobe was designed by combining hydrophobic rare earth nanoparticles with biocompatible mPEG-PLGA nanomicelles for near infrared II (NIR-II) imaging-guided anti-tumor chemotherapy. The as-synthesized nanoprobes (about 300 nm) with a highly enhanced permeability and retention (EPR) effect show great potential in the diagnosis of solid tumors, providing new prospects for clinical tumor diagnosis. Then, the degradable composite probes increase the imaging sensitivity of the probe and allow for the slow release of the internal anti-tumor drugs, reducing the loss of the drug during delivery. Finally, ultra-small rare earth nanoparticles (about 6 nm) can be excreted after hydrolysis of the composite probe to reduce the enrichment of the inorganic nanoparticles in vivo. Thus, this degradable NIR-II imaging probe based on a polymer-lanthanide composite could be a promising candidate for preclinical cancer chemotherapy and surgery navigation under a single 808 nm laser.
Sigma-Aldrich
Sodium trifluoroacetate, 98%