MilliporeSigma
  • CLIC4 is a cytokinetic cleavage furrow protein that regulates cortical cytoskeleton stability during cell division.
CLIC4 is a cytokinetic cleavage furrow protein that regulates cortical cytoskeleton stability during cell division.
Journal of cell science (2020-03-19)
Eric Peterman, Mindaugas Valius, Rytis Prekeris
ABSTRACT
During mitotic cell division, the actomyosin cytoskeleton undergoes several dynamic changes that play key roles in progression through mitosis. Although the regulators of cytokinetic ring formation and contraction are well established, proteins that regulate cortical stability during anaphase and telophase have been understudied. Here, we describe a role for CLIC4 in regulating actin and actin regulators at the cortex and cytokinetic cleavage furrow during cytokinesis. We first describe CLIC4 as a new component of the cytokinetic cleavage furrow that is required for successful completion of mitotic cell division. We also demonstrate that CLIC4 regulates the remodeling of the sub-plasma-membrane actomyosin network within the furrow by recruiting MST4 kinase (also known as STK26) and regulating ezrin phosphorylation. This work identifies and characterizes new molecular players involved in regulating cortex stiffness and blebbing during the late stages of cytokinetic furrowing.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Anillin Antibody, clone 5f3.1, clone 5F3.1, from mouse