MilliporeSigma
  • Copolymacrolactones Grafted with l-Glutamic Acid: Synthesis, Structure, and Nanocarrier Properties.
Copolymacrolactones Grafted with l-Glutamic Acid: Synthesis, Structure, and Nanocarrier Properties.
Polymers (2020-04-30)
Ernesto Tinajero-Díaz, Antxon Martínez de Ilarduya, Sebastián Muñoz-Guerra
ABSTRACT
The enzymatic ring-opening copolymerization (eROP) of globalide (Gl) and pentadecalactone (PDL) was performed in solution from mixtures of the two macrolactones at ratios covering the whole range of comonomeric compositions. The resulting P(Glx-r-PDLy) random copolyesters were aminofunctionalized by thiol-ene reaction with aminoethanethiol. ROP of γ-benzyl-l-glutamate N-carboxyanhydride initiated by P(Glx-r-PDLy)-NH2 provided neutral poly(γ-benzyl-l-glutamate)-grafted copolyesters, which were converted by hydrolysis into negatively charged hybrid copolymers. Both water-soluble and nonsoluble copolymers were produced depending on copolymer charge and their grafting degree, and their capacity for self-assembling in nano-objects were comparatively examined. The emulsion solvent-evaporation technique applied to the chloroform-soluble copolymers grafted with benzyl glutamate rendered well-delineated spherical nanoparticles with an average diameter of 200-300 nm. Conversely, micellar solutions in water were produced from copolyesters bearing grafted chains composed of at least 10 units of glutamic acid in the free form. The copolymer micelles were shown to be able to load doxorubicin (DOX) efficiently through electrostatic interactions and also to release the drug at a rate that was markedly pH dependent.
Sigma-Aldrich
2-(Boc-amino)ethanethiol, 97%
Sigma-Aldrich
Sodium trifluoroacetate, 98%