MilliporeSigma
  • Osteogenic impact of pro-apoptotic caspase inhibitors in MC3T3-E1 cells.
Osteogenic impact of pro-apoptotic caspase inhibitors in MC3T3-E1 cells.
Scientific reports (2020-05-06)
Adéla Kratochvílová, Barbora Veselá, Vojtěch Ledvina, Eva Švandová, Karel Klepárník, Kateřina Dadáková, Petr Beneš, Eva Matalová
ABSTRACT
Caspases are proteases traditionally associated with inflammation and cell death. Recently, they have also been shown to modulate cell proliferation and differentiation. The aim of the current research was to search for osteogenic molecules affected by caspase inhibition and to specify the individual caspases critical for these effects with a focus on proapoptotic caspases: caspase-2, -3, -6, -7, -8 and -9. Along with osteocalcin (Ocn), general caspase inhibition significantly decreased the expression of the Phex gene in differentiated MC3T3-E1 cells. The inhibition of individual caspases indicated that caspase-8 is a major contributor to the modification of Ocn and Phex expression. Caspase-2 and-6 had effects on Ocn and caspase-6 had an effect on Phex. These data confirm and expand the current knowledge about the nonapoptotic roles of caspases and the effect of their pharmacological inhibition on the osteogenic potential of osteoblastic cells.
Sigma-Aldrich
Fast Blue BB Salt hemi(zinc chloride) salt, Dye content ≥80 %
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Naphthol AS-TR phosphate disodium salt, ≥99% (HPLC), Bulk package