MilliporeSigma
  • Synthesis and Biological Evaluation of Three New Chitosan Schiff Base Derivatives.
Synthesis and Biological Evaluation of Three New Chitosan Schiff Base Derivatives.
ACS omega (2020-06-23)
Nadia Q Haj, Mohsin O Mohammed, Luqman E Mohammood
ABSTRACT
Recently, chemical modifications of chitosan (CS) have attracted the attention of scientific researchers due to its wide range of applications. In this research, chitin (CH) was extracted from the scales of Cyprinus carpio fish and converted to CS by three chemical steps: (i) demineralization, (ii) deprotonation, and (iii) deacetylation. The degree (measured as a percentage) of deacetylation (DD %) was calculated utilizing the acid-base titration method. The structure of CS was characterized by Fourier transform infrared (FT-IR) spectroscopy and thermogravimetric analysis (TGA). Three new CS Schiff bases (CSSBs) (CS-P1, CS-P2, and CS-P3) were synthesized via coupling of CS with 2-chloroquinoline-3-carbaldehyde, quinazoline-6-carbaldehyde, and oxazole-4-carbaldehyde, respectively. The newly prepared derivatives were verified, structurally, by nuclear magnetic resonance (1H and 13C NMR) and FT-IR spectroscopy. Antimicrobial activity was evaluated for the prepared compounds against both "Gram-negative" and "Gram-positive" bacteria, namely, Escherichia coli, Klebsiella pneumonia, Staphylococcus aureus, and Streptococcus mutans, in addition to two kinds of fungi, Candida albicans and Aspergillus fumigates. Cytotoxicity of the synthesized CSSBs was evaluated via a MTT screening test. The results indicated a critical activity increase of the synthesized compound rather than CS generally tested bacteria and fungi and the absence of cytotoxic activity. These findings suggested that these new CSSBs are novel biomaterial candidates with enhanced antibacterial and nontoxic characteristics for applications in areas of both biology and medicine.
Sigma-Aldrich
4-Hydroxyquinazoline, 98%
Sigma-Aldrich
4-Oxazolecarboxaldehyde, 97%