MilliporeSigma
  • Fluorescent Submicron-Sized Poly(heptafluoro-n-butyl methacrylate) Particles with Long-Term Stability.
Fluorescent Submicron-Sized Poly(heptafluoro-n-butyl methacrylate) Particles with Long-Term Stability.
Molecules (Basel, Switzerland) (2020-04-30)
Maciej Jarzębski, Przemysław Siejak, Monika Przeor, Jacek Gapiński, Anna Woźniak, Hanna Maria Baranowska, Jarosław Pawlicz, Elżbieta Baryła-Pankiewicz, Anna Szwajca
ABSTRACT
Fluorescent submicron particles of fluorinated methacrylate (HFMBA) with long-term stability have been synthesized and characterized with regard to their potential applications. Rhodamine B (RBITC) isothiocyanate was used as the fluorescent component. The core-shell structure of the particles effectively protected the dye against bleaching. HFBMA nanoparticle (NP) stability was confirmed after seven years of storage. Only slight differences were found in the polydispersity index (pdi) from 0.002 to 0.010. Particle size measurements were carried out using dynamic light scattering (DLS), nanoparticle tracking (NTA), and fluorescence correlation spectroscopy (FCS). The hydrodynamic diameter evaluated by different methods were in good agreement, respectively: 184-550 nm, 218-579 nm, and 236-508 nm. Particle and core morphology was estimated by using scanning and transmission electron microscopy (SEM and TEM). The ability to recognize particles in 3D as a reference sample in biological media has been confirmed by epifluorescence optical microscopy, confocal laser scanning microscopy, and super-resolution confocal microscopy (STED).
Sigma-Aldrich
2,2,3,3,4,4,4-Heptafluorobutyl methacrylate, contains MEHQ as inhibitor, 97%