MilliporeSigma
  • Peroxisome proliferator-activated receptor β/δ and γ agonists differentially affect prostaglandin E2 and cytokine synthesis and nutrient transporter expression in porcine trophoblast cells during implantation.
Peroxisome proliferator-activated receptor β/δ and γ agonists differentially affect prostaglandin E2 and cytokine synthesis and nutrient transporter expression in porcine trophoblast cells during implantation.
Theriogenology (2020-05-04)
Agnieszka Blitek, Magdalena Szymanska
ABSTRACT
Peroxisome proliferator-activated receptors (PPARs) belong to the nuclear receptor family of ligand-dependent transcription factors. PPARs have been shown to be important regulators of female reproductive functions, including conceptus development and placenta formation. This study examines the effect of PPARβ/δ and PPARγ agonists and antagonists on (1) the synthesis of prostaglandin (PG) E2, interleukin (IL) 6, interferon (IFN) γ, and tumor necrosis factor (TNF) α and (2) the mRNA expression of genes encoding nutrient transporters and/or binding proteins in Day 15 conceptus trophoblast cells. The study also examines whether PPAR agonist-modulated IL6, IFNγ, and TNFα secretion is mediated via mitogen-activated protein kinase (MAPK) pathways. Trophoblast cells were exposed to L-165,041 (a PPARβ/δ agonist) or rosiglitazone (a PPARγ agonist) in the presence or absence of GSK3787 (a PPARβ/δ antagonist) or GW9662 (a PPARγ antagonist) or in the presence or absence of U0126 (a MAPK inhibitor). Rosiglitazone stimulated PGE synthase and IFNG mRNA expression in trophoblast cells and enhanced PGE2 concentrations in the incubation medium. Moreover, cells treated with rosiglitazone exhibited increased abundance of the solute carrier organic anion transporter family member 2A1 (SLCO2A1, a PG transporter) and of fatty acid binding protein (FABP) 5 transcripts. All these effects were abolished by the addition of GW9662, which indicates that the action of rosiglitazone is PPARγ-dependent in the studied cells. L-165,041 inhibited TNFα synthesis and decreased the mRNA expression of FABP3 and IL6 in trophoblast cells. However, this effect was not abolished by the addition of GSK3787 into the incubation medium, suggesting that L-165,041 action is independent of PPARβ/δ. The inhibitory effect of L-165,041 on TNFα concentration and the stimulatory effect of rosiglitazone on IFNγ accumulation in the medium were not observed in the presence of the MAPK inhibitor, suggesting that the action of both agonists may be mediated by MAPKs. In conclusion, PPARβ/δ and PPARγ agonists are differentially involved in the trophoblast expression of genes related to conceptus development and implantation in pigs. Furthermore, L-165,041 and rosiglitazone may have PPAR-dependent and -independent effects in conceptus trophoblast cells.
Sigma-Aldrich
Newborn Calf Serum, USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
N-Chlorosuccinimide, 98%