MilliporeSigma
  • Effect of the level of "Candidatus Liberibacter solanacearum" infection on the development of zebra chip disease in different potato genotypes at harvest and post storage.
Effect of the level of "Candidatus Liberibacter solanacearum" infection on the development of zebra chip disease in different potato genotypes at harvest and post storage.
PloS one (2020-04-29)
Regina K Cruzado, Mahnaz Rashidi, Nora Olsen, Richard G Novy, Erik J Wenninger, Nilsa A Bosque-Pérez, Alexander V Karasev, William J Price, Arash Rashed
ABSTRACT
Potato psyllid (Bactericera cockerelli Sulc)-transmitted "Candidatus Liberibacter solanacearum" (Lso) has been negatively impacting the potato industry in the United States as well as other potato-producing countries. Lso has been linked to a condition known as zebra chip (ZC) that affects yield and quality of potato tubers. Efforts to find sources of resistance to ZC have primarily focused on greenhouse evaluations based on a single inoculation time prior to harvest. Plant response to infection, however, could be influenced by the developmental stage of the host plant, and ZC may continue to develop after harvest. The objectives of this study were to quantify Lso inoculation success, Lso titer, ZC severity and Lso development during storage in eight potato genotypes. These evaluations were conducted on plants infested with Lso-positive psyllids at 77, 12, and 4 days before vine removal (DBVR). The evaluated genotypes were categorized according to their relative resistance to Lso and tolerance to ZC symptoms. Lso inoculation success in the genotype family A07781, derived from Solanum chacoense, was lower than that of the susceptible control ('Russet Burbank'). A07781-4LB and A07781-3LB genotypes were characterized relatively resistant to the pathogen and highly tolerant to ZC symptoms, while A07781-10LB was categorized as susceptible to Lso but relatively tolerant to symptom expression. In stored potatoes, increase in Lso concentrations was observed for all infestation times. However, significantly higher Lso titer was detected in tubers infested 12 DBVR and the effect was similar across genotypes. Overall, the A07781 family can be considered as a promising source of resistance or tolerance to ZC.
Product Description
Supelco
Spirotetramat, PESTANAL®, analytical standard