MilliporeSigma
Epigenetic inheritance of circadian period in clonal cells.
eLife (2020-05-28)
Yan Li, Yongli Shan, Gokhul Krishna Kilaru, Stefano Berto, Guang-Zhong Wang, Kimberly H Cox, Seung-Hee Yoo, Shuzhang Yang, Genevieve Konopka, Joseph S Takahashi
ABSTRACT
Circadian oscillations are generated via transcriptional-translational negative feedback loops. However, individual cells from fibroblast cell lines have heterogeneous rhythms, oscillating independently and with different period lengths. Here we showed that heterogeneity in circadian period is heritable and used a multi-omics approach to investigate underlying mechanisms. By examining large-scale phenotype-associated gene expression profiles in hundreds of mouse clonal cell lines, we identified and validated multiple novel candidate genes involved in circadian period determination in the absence of significant genomic variants. We also discovered differentially co-expressed gene networks that were functionally associated with period length. We further demonstrated that global differential DNA methylation bidirectionally regulated these same gene networks. Interestingly, we found that depletion of DNMT1 and DNMT3A had opposite effects on circadian period, suggesting non-redundant roles in circadian gene regulation. Together, our findings identify novel gene candidates involved in periodicity, and reveal DNA methylation as an important regulator of circadian periodicity.
Sigma-Aldrich
Zebularine, ≥98% (HPLC), solid