MilliporeSigma
  • Cytoplasmic PPARγ is a marker of poor prognosis in patients with Cox-1 negative primary breast cancers.
Cytoplasmic PPARγ is a marker of poor prognosis in patients with Cox-1 negative primary breast cancers.
Journal of translational medicine (2020-02-23)
Wanting Shao, Christina Kuhn, Doris Mayr, Nina Ditsch, Magdalena Kailuwait, Verena Wolf, Nadia Harbeck, Sven Mahner, Udo Jeschke, Vincent Cavaillès, Sophie Sixou
ABSTRACT
The aim of this study was to investigate the expression of the nuclear receptor PPARγ, together with that of the cyclooxygenases Cox-1 and Cox-2, in breast cancer (BC) tissues and to correlate the data with several clinicobiological parameters including patient survival. In a well characterized cohort of 308 primary BC, PPARγ, Cox-1 and Cox-2 cytoplasmic and nuclear expression were evaluated by immunohistochemistry. Correlations with clinicopathological and aggressiveness features were analyzed, as well as survival using Kaplan-Meier analysis. PPARγ was expressed in almost 58% of the samples with a predominant cytoplasmic location. Cox-1 and Cox-2 were exclusively cytoplasmic. Cytoplasmic PPARγ was inversely correlated with nuclear PPARγ and ER expression, but positively with Cox-1, Cox-2, and other high-risk markers of BC, e.g. HER2, CD133, and N-cadherin. Overall survival analysis demonstrated that cytoplasmic PPARγ had a strong correlation with poor survival in the whole cohort, and even stronger in the subgroup of patients with no Cox-1 expression where cytoplasmic PPARγ expression appeared as an independent marker of poor prognosis. In support of this cross-talk between PPARγ and Cox-1, we found that Cox-1 became a marker of good prognosis only when cytoplasmic PPARγ was expressed at high levels. Altogether, these data suggest that the relative expression of cytoplasmic PPARγ and Cox-1 may play an important role in oncogenesis and could be defined as a potential prognosis marker to identify specific high risk BC subgroups.
Sigma-Aldrich
Anti-COX2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Anti-PTGS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution