MilliporeSigma
  • Indomethacin promotes browning and brown adipogenesis in both murine and human fat cells.
Indomethacin promotes browning and brown adipogenesis in both murine and human fat cells.
Pharmacology research & perspectives (2020-05-21)
Haley Overby, Yang Yang, Xinyun Xu, Shu Wang, Ling Zhao
ABSTRACT
Indomethacin (Indo), a nonsteroidal antiinflammatory drug, has been shown to promote murine brown adipogenesis both in vitro and in vivo, possibly due to its peroxisome proliferator-activated receptor gamma (PPARγ)-agonist activities. However, it is unclear whether Indo induces browning of white adipocytes from both murine and human origins or induces human brown adipogenesis. To bridge the gap, this study investigated the effects of increasing concentrations of Indo on murine 3T3-L1, human primary subcutaneous white adipocytes (HPsubQ), and human brown (HBr) adipocytes. The results show that Indo dose-dependently enhanced 3T3-L1 adipocyte differentiation and upregulated both mRNA and protein expression of brown and beige adipocyte markers, while simultaneously suppressing white adipocyte-specific marker mRNA expression. mRNA and protein expression of mitochondrial biogenesis and structural genes were dose-dependently enhanced in Indo treated 3T3-L1 adipocytes. This was accompanied by augmented mitochondrial DNA, enhanced oxygen consumption, proton leak, and maximal and spare respiratory capacity. Dose-dependent transactivation of PPARγ confirmed Indo's PPARγ-agonist activity in 3T3-L1 cells. Knockdown of PPARγ significantly attenuated Indo's activities in selective browning genes, demonstrating PPARγ dependence of these effects. Moreover, Indo enhanced mRNA and protein expression of brown markers in HPsubQ adipocytes. Interestingly, Indo-induced differential effects on individual PPARγ isoforms with significant dose-dependent induction of PPARγ-2 and suppression of PPARγ-1 protein expression. Finally, Indo significantly promoted brown adipogenesis in HBr cells. Taken together, these results demonstrate Indo to be a potent thermogenic compound in both murine and human fat cells and may be explored as a therapeutic agent for obesity treatment and prevention.
Sigma-Aldrich
Anti-ADRB3 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-UCP-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PGC-1 Antibody, Chemicon®, from rabbit