MilliporeSigma
  • Smarcad1 mediates microbiota-induced inflammation in mouse and coordinates gene expression in the intestinal epithelium.
Smarcad1 mediates microbiota-induced inflammation in mouse and coordinates gene expression in the intestinal epithelium.
Genome biology (2020-03-13)
Juri Kazakevych, Jérémy Denizot, Anke Liebert, Mariana Portovedo, Mia Mosavie, Payal Jain, Claudia Stellato, Claire Fraser, Renan Oliveira Corrêa, Marina Célestine, Raphaël Mattiuz, Hanneke Okkenhaug, J Ross Miller, Marco Aurélio Ramirez Vinolo, Marc Veldhoen, Patrick Varga-Weisz
ABSTRACT
How intestinal epithelial cells interact with the microbiota and how this is regulated at the gene expression level are critical questions. Smarcad1 is a conserved chromatin remodeling factor with a poorly understood tissue function. As this factor is highly expressed in the stem and proliferative zones of the intestinal epithelium, we explore its role in this tissue. Specific deletion of Smarcad1 in the mouse intestinal epithelium leads to colitis resistance and substantial changes in gene expression, including a striking increase of expression of several genes linked to innate immunity. Absence of Smarcad1 leads to changes in chromatin accessibility and significant changes in histone H3K9me3 over many sites, including genes that are differentially regulated upon Smarcad1 deletion. We identify candidate members of the gut microbiome that elicit a Smarcad1-dependent colitis response, including members of the poorly understood TM7 phylum. Our study sheds light onto the role of the chromatin remodeling machinery in intestinal epithelial cells in the colitis response and shows how a highly conserved chromatin remodeling factor has a distinct role in anti-microbial defense. This work highlights the importance of the intestinal epithelium in the colitis response and the potential of microbial species as pharmacological and probiotic targets in the context of inflammatory diseases.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
Sigma-Aldrich
Dispase® II, protease
Sigma-Aldrich
Dextran sulfate sodium salt, Mr ~40,000
Sigma-Aldrich
Anti-SMARCAD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution