MilliporeSigma
  • Blocking TG2 attenuates bleomycin-induced pulmonary fibrosis in mice through inhibiting EMT.
Blocking TG2 attenuates bleomycin-induced pulmonary fibrosis in mice through inhibiting EMT.
Respiratory physiology & neurobiology (2020-02-02)
Kai Wang, Cuihua Zu, Yan Zhang, Xiaojing Wang, Xiang Huan, Liwei Wang
ABSTRACT
Epithelial-mesenchymal transformation (EMT) is a central mechanism for the occurrence and development of pulmonary fibrosis. Therefore, to identify the key target molecules regulating the EMT process is considered as an important direction for the prevention and treatment of pulmonary fibrosis. Transglutaminase 2 (TG2) has been recently found to play an important role in the regulation of inflammation and the generation of extracellular matrix. Here, our study focuses on the roles of TG2 in pulmonary fibrosis and EMT. at first, the expression of TG2 and the EMT-related markers like E-cadherin, Vimentin, and α-SMA were detected with Western Blotting, immunohistochemistry and other methods in the mice with pulmonary fibrosis induced by bleomycin. Further, MLE 12 cells were used to study the effects on EMT of the inhibition of TG2 in vitro. Finally, GK921, an inhibitor against TG2, was used to show its function in both prevention and treatment of pulmonary fibrosis induced by bleomycin in mice. bleomycin succeeded to induce pulmonary fibrosis in mice, with increased TG2 expression, EMT and Akt activation. Knock-down of TG2 by siRNA technique in MLE 12 cell (a mouse alveolar epithelial cell line) and GK921 (an inhibitor of TG2) all inhibited the EMT process, however SC79, an activator of Akt rescued above inhibition. Finally, GK921 alleviated pulmonary fibrosis in mice induced by bleomycin. Blocking TG2 reduces bleomycin-induced pulmonary fibrosis in mice via inhibiting EMT.
Product Description
Bleomycin sulfate, British Pharmacopoeia (BP) Reference Standard