MilliporeSigma
  • Clozapine Improves Behavioral and Biochemical Outcomes in a MK-801-Induced Mouse Model of Schizophrenia.
Clozapine Improves Behavioral and Biochemical Outcomes in a MK-801-Induced Mouse Model of Schizophrenia.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer (2020-06-02)
Syed Suhail Andrabi, Shruti Vishnoi, Riya Madan, Neha Bhardwaj, Heena Tabassum, Mohd Akram, Suhel Parvez
ABSTRACT
Glutamatergic N-methyl-D-aspartate (NMDA) receptors have critical roles in several neurological and psychiatric diseases. Dizocilpine (MK-801) is a ligand at phencyclidine recognition sites that is associated with NMDA receptor-coupled cation channels, where it acts as a potent noncompetitive antagonist of central glutamate receptors. In this study, we investigate the effect of clozapine on MK-801-induced neurochemical and neurobehavioral alterations in the prefrontal cortex of mice. Acute administration of NMDA noncompetitive antagonist MK-801 impairs motor coordination, grip strength, and locomotor activity. Clozapine is the only medication that is indicated for treating refractory schizophrenia, due to its superior efficacy among all antipsychotic agents; however, its mechanism is not well understood. To understand its mechanism, we investigated the effects of clozapine on motor coordination, locomotor activity, and grip strength in mice against the NMDA receptor antagonist MK-801. MK-801 induced elevations in acetylcholinesterase (AChE) activity, monoamine oxidase (MAO) activity, and c-fos expression. The administration of clozapine inhibited the effects caused by MK-801 (0.2 mg/kg body weight). Motor coordination and grip strength paradigms that had been altered by MK-801 were restored by clozapine. Moreover, clozapine also ameliorated MK-801-induced elevation in AChE and MAO activity. Our immunostaining results demonstrated that clozapine treatment reduced overexpression of the neuronal activity marker c-fos in cortices of the brain. Results of the current study determine that clozapine ameliorated cognition in MK-801-treated mice via cholinergic and neural mechanisms. These findings show that clozapine possesses the potential to augment cognition in diseases such as schizophrenia.
Sigma-Aldrich
Benzylamine hydrochloride