MilliporeSigma
  • Mitotic cell responses to substrate topological cues are independent of the molecular nature of adhesion.
Mitotic cell responses to substrate topological cues are independent of the molecular nature of adhesion.
Science signaling (2020-02-27)
Ouranio Anastasiou, Rania Hadjisavva, Paris A Skourides
ABSTRACT
Correct selection of the cell division axis is important for cell differentiation, tissue and organ morphogenesis, and homeostasis. Both integrins, which mediate interactions with extracellular matrix (ECM) components such as fibronectin, and cadherins, which mediate interactions between cells, are implicated in the determination of spindle orientation. We found that both cadherin- and integrin-based adhesion resulted in cell divisions parallel to the attachment plane and elicited identical spindle responses to spatial adhesive cues. This suggests that adhesion topology provides purely mechanical spatial cues that are independent of the molecular nature of the interaction or signaling from adhesion complexes. We also demonstrated that cortical integrin activation was indispensable for correct spindle orientation on both cadherin and fibronectin substrates. These data suggest that spindle orientation responses to adhesion topology are primarily a result of force anisotropy on the cell cortex and show that integrins play a central role in this process that is distinct from their role in cell-ECM interactions.
Sigma-Aldrich
Triethoxysilane, 95%
Sigma-Aldrich
Anti-LGN Antibody, serum, from rabbit
Sigma-Aldrich
PP2, ≥98% (HPLC)