MilliporeSigma
  • Autophagy promotes citrullination of VIM (vimentin) and its interaction with major histocompatibility complex class II in synovial fibroblasts.
Autophagy promotes citrullination of VIM (vimentin) and its interaction with major histocompatibility complex class II in synovial fibroblasts.
Autophagy (2019-09-06)
Eri Sugawara, Masaru Kato, Yuki Kudo, Wenshi Lee, Ryo Hisada, Yuichiro Fujieda, Kenji Oku, Toshiyuki Bohgaki, Olga Amengual, Shinsuke Yasuda, Tomohiro Onodera, Shigetsugu Hatakeyama, Tatsuya Atsumi
ABSTRACT
We aimed to investigate the involvement of macroautophagy/autophagy in autoimmunity in rheumatoid arthritis (RA) through citrullination of VIM (vimentin) and its interaction with MHC class II in synovial fibroblasts (SFs). The cell surface expression of MHC class II and B7 costimulatory molecules on SFs was analyzed by flow cytometry after treatment with IFNG/IFN-γ (interferon gamma). Intracellular citrullinated autoantigens in SFs were analyzed by immunoblotting using serum from anti-citrullinated peptide antibodies (ACPA)-positive patient as a primary antibody. SFs were incubated in serum-free medium or treated with proteasome inhibitor MG132 to induce autophagy. An autophagy inhibitor 3-methyladenin (3-MA) was used. Intracellular citrullinated VIM (cVIM) was evaluated by immunoblotting and immunocytochemistry. The interaction between MHC class II and cVIM was evaluated with co-immunoprecipitation and proximity ligation assay (PLA). We demonstrated that MHC class II, CD274/B7-H1 and PDCD1LG2/B7-DC were expressed on SFs following treatment with IFNG whereas CD276/B7-H3 was detected on SFs regardless of the presence of IFNG. ACPA-positive sera recognized a 54 kDa protein in SFs. By immunoprecipitation, the 54 kDa protein recognized by RA sera was revealed to be cVIM. Following induction of autophagy, intracellular cVIM was increased in SFs but the effect was canceled by 3-MA. The interaction between MHC class II and cVIM was demonstrated by co-immunoprecipitation. Furthermore, PLA revealed the significant increase of MHC class II-cVIM interaction following induction of autophagy. Our findings suggest that SFs may contribute to the autoimmunity in RA through citrullination of VIM and its interaction with MHC class II promoted by autophagy.Abbreviations: 3-MA: 3-methyladenine; ACPA: anti-citrullinated peptide antibodies; anti-CCP: anti-cyclic citrullinated peptide antibody; cVIM: citrullinated VIM; BECN1: beclin1; DAPI: 4',6-diamidino-2-phenylindole; FBS: fetal bovine serum; HLA: human leukocyte antigen; IFNG/IFN-γ: interferon gamma; IL6: interleukin 6; IP: immunoprecipitation; MAP1LC3/LC3: microtubule associated protein 1 light chain 3; MFI: mean fluorescence index; MHC: major histocompatibility complex; OA: osteoarthritis; PADI: peptidyl arginine deiminase; PepA: pepstatin A; PBS: phosphate-buffered saline; PtdIns3K: phosphatidylinositol 3-kinase; RA: rheumatoid arthritis; SFs: synovial fibroblasts; siRNA: small interfering RNA; VIM: vimentin.
Product Description
SAFC
Iscove′s Modified Dulbecco′s Medium, With sodium bicarbonate and L-glutamine, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization.
Sigma-Aldrich
Anti-Citrulline Antibody, Upstate®, from rabbit
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Millipore
Immobilon®-P PVDF Membrane, 10 sheets, 26 cm x 26 cm, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.