MilliporeSigma
  • High-resolution optoacoustic imaging of tissue responses to vascular-targeted therapies.
High-resolution optoacoustic imaging of tissue responses to vascular-targeted therapies.
Nature biomedical engineering (2020-03-14)
Katja Haedicke, Lilach Agemy, Murad Omar, Andrei Berezhnoi, Sheryl Roberts, Camila Longo-Machado, Magdalena Skubal, Karan Nagar, Hsiao-Ting Hsu, Kwanghee Kim, Thomas Reiner, Jonathan Coleman, Vasilis Ntziachristos, Avigdor Scherz, Jan Grimm
ABSTRACT
The monitoring of vascular-targeted therapies using magnetic resonance imaging, computed tomography or ultrasound is limited by their insufficient spatial resolution. Here, by taking advantage of the intrinsic optical properties of haemoglobin, we show that raster-scanning optoacoustic mesoscopy (RSOM) provides high-resolution images of the tumour vasculature and of the surrounding tissue, and that the detection of a wide range of ultrasound bandwidths enables the distinction of vessels of differing size, providing detailed insights into the vascular responses to vascular-targeted therapy. Using RSOM to examine the responses to vascular-targeted photodynamic therapy in mice with subcutaneous xenografts, we observed a substantial and immediate occlusion of the tumour vessels followed by haemorrhage within the tissue and the eventual collapse of the entire vasculature. Using dual-wavelength RSOM, which distinguishes oxyhaemoglobin from deoxyhaemoglobin, we observed an increase in oxygenation of the entire tumour volume immediately after the application of the therapy, and a second wave of oxygen reperfusion approximately 24 h thereafter. We also show that RSOM enables the quantification of differences in neoangiogenesis that predict treatment efficacy.
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Eosin Y Solution, Aqueous