MilliporeSigma
  • Antifungal Activity of an Original Amino-Isocyanonaphthalene (ICAN) Compound Family: Promising Broad Spectrum Antifungals.
Antifungal Activity of an Original Amino-Isocyanonaphthalene (ICAN) Compound Family: Promising Broad Spectrum Antifungals.
Molecules (Basel, Switzerland) (2020-02-23)
Miklós Nagy, Gábor Szemán-Nagy, Alexandra Kiss, Zsolt László Nagy, László Tálas, Dávid Rácz, László Majoros, Zoltán Tóth, Zsuzsa Máthéné Szigeti, István Pócsi, Sándor Kéki
ABSTRACT
: Multiple drug resistant fungi pose a serious threat to human health, therefore the development of completely new antimycotics is of paramount importance. The in vitro antifungal activity of the original, 1-amino-5-isocyanonaphthalenes (ICANs) was evaluated against reference strains of clinically important Candida species. Structure-activity studies revealed that the naphthalene core and the isocyano- together with the amino moieties are all necessary to exert antifungal activity. 1,1-N-dimethylamino-5-isocyanonaphthalene (DIMICAN), the most promising candidate, was tested further in vitro against clinical isolates of Candida species, yielding a minimum inhibitory concentration (MIC) of 0.04-1.25 µg/mL. DIMICAN was found to be effective against intrinsically fluconazole resistant Candidakrusei isolates, too. In vivo experiments were performed in a severly neutropenic murine model inoculated with a clinical strain of Candidaalbicans. Daily administration of 5 mg/kg DIMICAN intraperitoneally resulted in 80% survival even at day 13, whereas 100% of the control group died within six days. Based on these results, ICANs may become an effective clinical lead compound family against fungal pathogens.
Sigma-Aldrich
Iodoethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
1-Iodopropane, 99%
Sigma-Aldrich
1,5-Diaminonaphthalene, 97%