MilliporeSigma
Role of DJ-1 in Modulating Glycative Stress in Heart Failure.
Journal of the American Heart Association (2020-02-19)
Yuuki Shimizu, Chad K Nicholson, Rohini Polavarapu, Yvanna Pantner, Ahsan Husain, Nawazish Naqvi, Lih-Shen Chin, Lian Li, John W Calvert
ABSTRACT
Background DJ-1 is a ubiquitously expressed protein typically associated with the development of early onset Parkinson disease. Recent data suggest that it also plays a role in the cellular response to stress. Here, we sought to determine the role DJ-1 plays in the development of heart failure. Methods and Results Initial studies found that DJ-1 deficient mice (DJ-1 knockout; male; 8-10 weeks of age) exhibited more severe left ventricular cavity dilatation, cardiac dysfunction, hypertrophy, and fibrosis in the setting of ischemia-reperfusion-induced heart failure when compared with wild-type littermates. In contrast, the overexpression of the active form of DJ-1 using a viral vector approach resulted in significant improvements in the severity of heart failure when compared with mice treated with a control virus. Subsequent studies aimed at evaluating the underlying protective mechanisms found that cardiac DJ-1 reduces the accumulation of advanced glycation end products and activation of the receptor for advanced glycation end products-thus, reducing glycative stress. Conclusions These results indicate that DJ-1 is an endogenous cytoprotective protein that protects against the development of ischemia-reperfusion-induced heart failure by reducing glycative stress. Our findings also demonstrate the feasibility of using a gene therapy approach to deliver the active form of DJ-1 to the heart as a therapeutic strategy to protect against the consequences of ischemic injury, which is a major cause of death in western populations.
Sigma-Aldrich
Rat RAGE / AGER ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse RAGE / AGER ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Glyoxalase I Activity Assay Kit, sufficient for 100 tests (UV)