MilliporeSigma
  • Prenatal and postnatal alcohol exposure increases vulnerability to cocaine addiction in adult mice.
Prenatal and postnatal alcohol exposure increases vulnerability to cocaine addiction in adult mice.
British journal of pharmacology (2019-11-11)
Lídia Cantacorps, Sandra Montagud-Romero, Miguel Ángel Luján, Olga Valverde
ABSTRACT
Alcohol exposure in utero may lead to a wide range of long-lasting morphological and behavioural deficiencies known as fetal alcohol spectrum disorders (FASD), associated with a higher risk of later developing neuropsychiatric disorders. However, little is known about the long-term consequences of cocaine use and abuse in individuals with FASD. This study aimed to investigate the effects of maternal binge alcohol drinking during prenatal and postnatal periods on cocaine reward-related behaviours in adult offspring. Pregnant C57BL/6 female mice were exposed to an experimental protocol of binge alcohol consumption (drinking-in-the-dark test) from gestation to weaning. Male offspring were subsequently left undisturbed until reaching adulthood and were tested for cocaine-induced motivational responses (conditioned place preference, behavioural sensitization and operant self-administration). Protein expression of dopamine- and glutamate-related molecules was assessed following cocaine-induced reinstatement. The results show that prenatal and postnatal alcohol exposure enhanced the preference for the cocaine-paired chamber in the conditioned place preference test. Furthermore, early alcohol-exposed mice displayed attenuated cocaine-induced behavioural sensitization but also higher cocaine self-administration. Furthermore, alterations in glutamatergic excitability (GluA1/GluA2 ratio) and ΔFosB expression were found in the prefrontal cortex and the striatum of alcohol-exposed mice after cocaine-primed reinstatement. Our findings demonstrate that maternal binge-like alcohol consumption during gestation and lactation alters sensitivity to the reinforcing effects of cocaine in adult offspring mice. Together, such data suggest that prenatal and postnatal alcohol exposure may underlie an enhanced susceptibility of alcohol-exposed offspring to develop drug addiction later in adulthood.
Sigma-Aldrich
Anti-DARPP-32 Antibody, phosphoThr34, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GluR1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-GluR2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho-CREB (Ser133) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Dopamine D₂ Receptor Rabbit pAb, lyophilized, Calbiochem®
Sigma-Aldrich
Cocaine free base
Sigma-Aldrich
Anti-CREB Antibody, clone NL904, rabbit monoclonal, culture supernatant, clone NL904, Upstate®