MilliporeSigma
  • Alloxan Disintegrates the Plant Cytoskeleton and Suppresses mlo-Mediated Powdery Mildew Resistance.
Alloxan Disintegrates the Plant Cytoskeleton and Suppresses mlo-Mediated Powdery Mildew Resistance.
Plant & cell physiology (2019-11-19)
Hongpo Wu, Weiwei Zhang, Martin Schuster, Marcin Moch, Reinhard Windoffer, Gero Steinberg, Christopher J Staiger, Ralph Panstruga
ABSTRACT
Recessively inherited mutant alleles of Mlo genes (mlo) confer broad-spectrum penetration resistance to powdery mildew pathogens in angiosperm plants. Although a few components are known to be required for mlo resistance, the detailed molecular mechanism underlying this type of immunity remains elusive. In this study, we identified alloxan (5,5-dihydroxyl pyrimidine-2,4,6-trione) and some of its structural analogs as chemical suppressors of mlo-mediated resistance in monocotyledonous barley (Hordeum vulgare) and dicotyledonous Arabidopsis thaliana. Apart from mlo resistance, alloxan impairs nonhost resistance in Arabidopsis. Histological analysis revealed that the chemical reduces callose deposition and hydrogen peroxide accumulation at attempted fungal penetration sites. Fluorescence microscopy revealed that alloxan interferes with the motility of cellular organelles (peroxisomes, endosomes and the endoplasmic reticulum) and the pathogen-triggered redistribution of the PEN1/SYP121 t-SNARE protein. These cellular defects are likely the consequence of disassembly of actin filaments and microtubules upon alloxan treatment. Similar to the situation in animal cells, alloxan elicited the temporary accumulation of reactive oxygen species (ROS) in cotyledons and rosette leaves of Arabidopsis plants. Our results suggest that alloxan may destabilize cytoskeletal architecture via induction of an early transient ROS burst, further leading to the failure of molecular and cellular processes that are critical for plant immunity.
Sigma-Aldrich
2-Hydroxypyrimidine hydrochloride, 98%
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Barbituric acid, ReagentPlus®, 99%
Sigma-Aldrich
Alloxan monohydrate, 98%