MilliporeSigma
  • Proteomic mechanisms for the stimulatory effects of antibiotics on Microcystis aeruginosa during hydrogen peroxide treatment.
Proteomic mechanisms for the stimulatory effects of antibiotics on Microcystis aeruginosa during hydrogen peroxide treatment.
Chemosphere (2020-01-14)
Ying Liu, Jian Zhang, Baoyu Gao
ABSTRACT
Application of low dosage of H2O2 at early stage of cyanobacterial life cycle is a promising route for cyanobacterial bloom mitigation, which could minimize adverse effects on non-target organisms. Besides, influence of co-existing contaminants on cyanobacterial bloom mitigation under combined pollution conditions remains unclear. This study assessed the influence of a mixture of four frequently detected antibiotics (tetracycline, sulfamethoxazole, ciprofloxacin and amoxicillin) during H2O2 treatment of Microcystis aeruginosa at early growth stage. H2O2 significantly (p < 0.05) inhibited growth rate, chlorophyll a content, Fv/Fm and rETRmax in a dose-dependent manner at low doses of 0.25-1 mg L-1, through downregulating proteins involved in cell division, cellular component organization, gene expression and photosynthesis. Although H2O2 increased microcystin content in each cyanobacterial cell through the upregulation of microcystin synthetases (mcyC and mcyF), total microcystin concentration in H2O2 treated groups was significantly (p < 0.05) reduced due to the decrease of cell density. Existence of 80 and 200 ng L-1 mixed antibiotics during H2O2 treatment facilitated the scavenging of ROS by antioxidant enzymes and significantly (p < 0.05) stimulated growth, photosynthesis, microcystin synthesis and microcystin release in H2O2 treated cells, through the upregulation of proteins involved in photosynthesis, oxidation-reduction process, biosynthesis, gene expression and transport. Mixed antibiotics increased the hazard of M. aeruginosa during H2O2 treatment, through the stimulation of microcystin synthesis and release at the proteomic level. Each target antibiotic should be controlled below 5 ng L-1 before the application of H2O2 for eliminating the interference of antibiotics on cyanobacterial bloom mitigation.
Sigma-Aldrich
4-Nitrophenylboronic acid, ≥95.0%