MilliporeSigma
  • Bacteriostatic Effect of Piezoelectric Poly-3-Hydroxybutyrate and Polyvinylidene Fluoride Polymer Films under Ultrasound Treatment.
Bacteriostatic Effect of Piezoelectric Poly-3-Hydroxybutyrate and Polyvinylidene Fluoride Polymer Films under Ultrasound Treatment.
Polymers (2020-01-24)
Ivan S Vatlin, Roman V Chernozem, Alexander S Timin, Anna P Chernova, Evgeny V Plotnikov, Yulia R Mukhortova, Maria A Surmeneva, Roman A Surmenev
ABSTRACT
Antibiotic resistance of bacteria stimulates the development of new treatment approaches. Piezoelectric-catalysis has attracted much attention due to the possibility to effectively provide antibacterial effect via generation of reactive oxygen species. However, the influence of the surface charge or potential of a piezopolymer on bacteria has not been sufficiently studied so far. This study reports the fabrication and characterization of thin films of piezoelectric polyhydroxybutyrate, polyvinylidene fluoride, and polyvinylidene fluoride trifluoroethylene as well as non-piezoelectric polycaprolactone polymers fabricated using solution casting approach. The piezoelectric coefficient (d33) and surface electric peak-to-peak potential generated by the cyclic mechanical stress applied to the films were measured. Neither any toxic effect of the polymer films nor ultrasound influence on Escherichia coli bacteria behavior is observed. However, significant inhibition of the growth of bacteria is revealed during mechanical stimulation of piezoelectric samples via ultrasound treatment. Thus, this study demonstrates clear bacteriostatic effect of piezoelectric polymers for different tissue engineering applications.
Sigma-Aldrich
Trifluoroethylene, ≥97.5% (GC)