MilliporeSigma
  • Highly Deformable and See-Through Polymer Light-Emitting Diodes with All-Conducting-Polymer Electrodes.
Highly Deformable and See-Through Polymer Light-Emitting Diodes with All-Conducting-Polymer Electrodes.
Advanced materials (Deerfield Beach, Fla.) (2017-12-07)
Seyoung Kee, Nara Kim, Byoungwook Park, Bong Seong Kim, Soonil Hong, Jong-Hoon Lee, Soyeong Jeong, Ahryun Kim, Soo-Young Jang, Kwanghee Lee
ABSTRACT
Despite the high expectation of deformable and see-through displays for future ubiquitous society, current light-emitting diodes (LEDs) fail to meet the desired mechanical and optical properties, mainly because of the fragile transparent conducting oxides and opaque metal electrodes. Here, by introducing a highly conductive nanofibrillated conducting polymer (CP) as both deformable transparent anode and cathode, ultraflexible and see-through polymer LEDs (PLEDs) are demonstrated. The CP-based PLEDs exhibit outstanding dual-side light-outcoupling performance with a high optical transmittance of 75% at a wavelength of 550 nm and with an excellent mechanical durability of 9% bending strain. Moreover, the CP-based PLEDs fabricated on 4 µm thick plastic foils with all-solution processing have extremely deformable and foldable light-emitting functionality. This approach is expected to open a new avenue for developing wearable and attachable transparent displays.
Sigma-Aldrich
1-Ethyl-3-methylimidazolium tetracyanoborate Solarpur®